Assay Design



Gene Transcripts

Gene Symbol
Zfp52
Entrez Gene ID
22710
Gene Name
zinc finger protein 52
Gene Aliases
KRAB11, Zfp-52, Zfp76, mszf23-2, mszf24, mszf34, mszf75, zfec29
Chromosome Location
Chr.17: 21516295 - 21562601 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144515.2 NP_653098.2 1 - 2 298 60
GenBank mRNA AK032299.1 - 1 - 2 299 60
BC021527.1 - 1 - 2 210 60

View Full Product Details