Assay Design



Gene Transcripts

Gene Symbol
Mlf2
Entrez Gene ID
30853
Gene Name
myeloid leukemia factor 2
Gene Aliases
AI256856, AU018944, C77284
Chromosome Location
Chr.6: 124931388 - 124936149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145385.2 NP_663360.1 8 - 9 998 51
GenBank mRNA BC003975.1 - 8 - 9 909 51
CD560861.1 - 1 - 2 112 51

View Full Product Details