Assay Design



Gene Transcripts

Gene Symbol
Phyhip
Entrez Gene ID
105653
Gene Name
phytanoyl-CoA hydroxylase interacting protein
Gene Aliases
AW049870, C630010D02Rik, Lnap1ip, PAHX-AP#1, PAHX-AP1
Chromosome Location
Chr.14: 70457517 - 70468826 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145981.3 NP_666093.1 4 - 5 763 81
GenBank mRNA AK049900.1 - 4 - 5 764 81
BC030494.1 - 4 - 5 760 81
BC139360.1 - 5 - 6 746 81

View Full Product Details