Assay Design



Gene Transcripts

Gene Symbol
Slc5a11
Entrez Gene ID
233836
Gene Name
solute carrier family 5 (sodium/glucose cotransporter), member 11
Gene Aliases
2010013B02Rik, Kst1, RKST2, SMIT2, Slc5a10
Chromosome Location
Chr.7: 123214758 - 123274390 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146198.2 NP_666310.1 16 - 17 2088 66
GenBank mRNA AK008219.1 - 16 - 17 2143 66
BC031742.1 - 16 - 17 2089 66

View Full Product Details