Assay Design



Gene Transcripts

Gene Symbol
Leap2
Entrez Gene ID
259301
Gene Name
liver-expressed antimicrobial peptide 2
Gene Aliases
leap-2
Chromosome Location
Chr.11: 53422181 - 53423136 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153069.3 NP_694709.1 1 - 2 86 77
GenBank mRNA AJ409055.1 - 1 - 2 86 77
BC089593.1 - 1 - 2 79 77
W82161.1 - 1 - 2 120 77

View Full Product Details