Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253702.1 | NP_001240631.1 | 12 - 13 | 1900 | 96 |
NM_001253703.1 | NP_001240632.1 | 10 - 11 | 1762 | 96 | |
NM_153091.3 | NP_694731.1 | 12 - 13 | 1996 | 96 | |
NR_045578.1 | 12 - 13 | 2262 | 96 | ||
GenBank mRNA | AB084518.1 | - | 12 - 13 | 1410 | 96 |
AK033865.1 | - | 12 - 13 | 1427 | 96 | |
AK144075.1 | - | 10 - 11 | 1202 | 96 | |
AK148099.1 | - | 12 - 13 | 1696 | 96 | |
AK172102.1 | - | 12 - 13 | 1901 | 96 | |
AK172218.1 | - | 12 - 13 | 1901 | 96 | |
BC024473.1 | - | 12 - 13 | 1387 | 96 | |
BC058781.1 | - | 12 - 13 | 1421 | 96 | |
BC107394.2 | - | 12 - 13 | 1421 | 96 | |
BC145950.1 | - | 12 - 13 | 1488 | 96 |