Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153122.2 | NP_694762.1 | 14 - 15 | 2055 | 70 |
GenBank mRNA | AK158391.1 | - | 14 - 15 | 2097 | 70 |
BC025120.1 | - | 1 - 2 | 153 | 70 | |
BC034522.1 | - | 14 - 15 | 2055 | 70 |