Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290824.1 | NP_001277753.1 | 1 - 2 | 309 | 101 |
NM_153387.3 | NP_700436.1 | 1 - 2 | 309 | 101 | |
NR_077245.2 | 1 - 2 | 309 | 101 | ||
NR_110985.1 | 1 - 2 | 309 | 101 | ||
GenBank mRNA | AK038921.1 | - | 1 - 2 | 268 | 101 |
AK045305.1 | - | 1 - 2 | 202 | 101 | |
AK046132.1 | - | 1 - 2 | 239 | 101 | |
AK050250.1 | - | 1 - 2 | 310 | 101 | |
AK051517.1 | - | 1 - 2 | 210 | 101 | |
AK152984.1 | - | 1 - 2 | 268 | 101 | |
AK153171.1 | - | 1 - 2 | 268 | 101 | |
BC029106.1 | - | 1 - 2 | 273 | 101 |