Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242430.1 | NP_001229359.1 | 2 - 3 | 688 | 78 |
NM_153515.4 | NP_705735.1 | 2 - 3 | 487 | 78 | |
GenBank mRNA | AK030009.1 | - | 2 - 3 | 695 | 78 |
AK054131.1 | - | 2 - 3 | 486 | 78 | |
AK133018.1 | - | 2 - 3 | 689 | 78 | |
AK151060.1 | - | 2 - 3 | 488 | 78 | |
AK151257.1 | - | 2 - 3 | 388 | 78 | |
BC012310.1 | - | 1 - 2 | 92 | 78 | |
BC034661.1 | - | 2 - 3 | 228 | 78 | |
BC056948.1 | - | 2 - 3 | 431 | 78 |