Assay Design



Gene Transcripts

Gene Symbol
Rell2
Entrez Gene ID
225392
Gene Name
RELT-like 2
Gene Aliases
4631403P03Rik, ependolin
Chromosome Location
Chr.18: 37955084 - 37959179 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153793.2 NP_722488.2 1 - 2 354 53
GenBank mRNA AK044097.1 - 1 - 2 317 53
BC038500.1 - 1 - 2 297 53
BY262749.1 - 1 - 2 354 53

View Full Product Details