Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153799.3 | NP_722494.1 | 6 - 7 | 1336 | 55 |
GenBank mRNA | AK090005.1 | - | - | 1027 | 55 |
AK140127.1 | - | 5 - 6 | 1031 | 55 | |
AK146804.1 | - | 6 - 7 | 1366 | 55 | |
AK166286.1 | - | 6 - 7 | 1333 | 55 | |
AK168118.1 | - | 6 - 7 | 1344 | 55 | |
BC031725.1 | - | 6 - 7 | 1336 | 55 | |
BC033484.1 | - | 6 - 7 | 1337 | 55 |