Assay Design
Gene Transcripts
- Gene Symbol
- Chil5
- Entrez Gene ID
-
229687
- Gene Name
- chitinase-like 5
- Gene Aliases
-
Bclp, Bclp1, Bclp2, Chi3l7
- Chromosome Location
-
Chr.3: 106016919 - 106033131 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB081756.1 |
- |
8 - 9 |
934 |
122 |
BC116403.1 |
- |
6 - 7 |
762 |
122 |
View Full Product Details