Assay Design



Gene Transcripts

Gene Symbol
Brca2
Entrez Gene ID
12190
Gene Name
breast cancer 2
Gene Aliases
Fancd1, RAB163
Chromosome Location
Chr.5: 150522021 - 150570147 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081001.2 NP_001074470.1 9 - 10 1277 121
NM_009765.3 NP_033895.2 9 - 10 1083 121
GenBank mRNA AK145017.1 - 9 - 10 1244 121
AK148272.1 - 9 - 10 1081 121
AK164598.1 - 7 - 8 1589 121
U65594.1 - 9 - 10 1060 121
U82270.1 - 8 - 9 769 121
U89652.1 - 8 - 9 807 121

View Full Product Details