Assay Design



Gene Transcripts

Gene Symbol
Emilin1
Entrez Gene ID
100952
Gene Name
elastin microfibril interfacer 1
Gene Aliases
5830419M17Rik, AW229038, BB105748, EMILIN-1, gp115
Chromosome Location
Chr.5: 30913381 - 30921279 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133918.2 NP_598679.1 6 - 7 2666 101
GenBank mRNA AK029337.1 - 6 - 7 2637 101
AK052368.1 - 6 - 7 2746 101
AK088995.1 - 6 - 7 2732 101
AK140388.1 - 4 - 5 2082 101
AK155486.1 - 6 - 7 2968 101
BC005481.1 - 6 - 7 2666 101

View Full Product Details