Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133918.2 | NP_598679.1 | 7 - 8 | 2805 | 99 |
GenBank mRNA | AK029337.1 | - | 7 - 8 | 2776 | 99 |
AK052368.1 | - | 7 - 8 | 2885 | 99 | |
AK088995.1 | - | 7 - 8 | 2871 | 99 | |
AK140388.1 | - | 5 - 6 | 2221 | 99 | |
AK155486.1 | - | 7 - 8 | 3107 | 99 | |
BC005481.1 | - | 7 - 8 | 2805 | 99 |