Assay Design



Gene Transcripts

Gene Symbol
Edem2
Entrez Gene ID
108687
Gene Name
ER degradation enhancer, mannosidase alpha-like 2
Gene Aliases
9530090G24Rik, AI327354
Chromosome Location
Chr.2: 155701673 - 155729475 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145537.2 NP_663512.2 10 - 11 1377 64
GenBank mRNA AK029164.1 - 10 - 11 1329 64
AK075650.1 - 10 - 11 1291 64
AK079296.1 - 10 - 11 1313 64
AK080636.1 - 10 - 11 1322 64
AK149892.1 - 10 - 11 1312 64
AK155024.1 - 10 - 11 1340 64
AK166998.1 - 10 - 11 1313 64
BC008268.1 - 10 - 11 1288 64

View Full Product Details