Assay Design



Gene Transcripts

Gene Symbol
Hes3
Entrez Gene ID
15207
Gene Name
hairy and enhancer of split 3 (Drosophila)
Gene Aliases
Hes-3, bHLHb43
Chromosome Location
Chr.4: 152285250 - 152291903 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008237.4 NP_032263.2 4 - 5 544 93
GenBank mRNA AK090218.1 - 4 - 5 545 93
BC116444.2 - 3 - 4 459 93

View Full Product Details