Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010404.3 | NP_034534.1 | 8 - 9 | 1189 | 76 |
NM_177981.2 | NP_817090.1 | 8 - 9 | 1189 | 76 | |
GenBank mRNA | AJ000262.1 | - | 8 - 9 | 1135 | 76 |
AJ002271.1 | - | 8 - 9 | 1135 | 76 | |
AK138436.1 | - | - | 1138 | 76 | |
BC034089.1 | - | 8 - 9 | 1127 | 76 | |
BC053043.1 | - | 8 - 9 | 1176 | 76 |