Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010431.2 | NP_034561.2 | 14 - 15 | 2782 | 103 |
GenBank mRNA | AF003695.1 | - | 14 - 15 | 2616 | 103 |
AK033471.1 | - | 3 - 4 | 382 | 103 | |
AK034087.1 | - | 14 - 15 | 2748 | 103 | |
AK076395.1 | - | 12 - 13 | 2070 | 103 | |
AK150367.1 | - | 14 - 15 | 2649 | 103 | |
BC026139.1 | - | 14 - 15 | 2634 | 103 | |
X95580.1 | - | 14 - 15 | 2406 | 103 |