Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012016.2 | NP_036146.2 | 20 - 21 | 2522 | 86 |
GenBank mRNA | AF071777.1 | - | 20 - 21 | 2481 | 86 |
AK153659.1 | - | 20 - 21 | 2512 | 86 | |
BC108999.2 | - | 20 - 21 | 2478 | 86 | |
BC109000.1 | - | 20 - 21 | 2478 | 86 |