Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252457.1 | NP_001239386.1 | 2 - 3 | 357 | 53 |
NM_019693.3 | NP_062667.1 | 2 - 3 | 445 | 53 | |
GenBank mRNA | AF118128.1 | - | 1 - 2 | 211 | 53 |
AK051034.1 | - | 2 - 3 | 378 | 53 | |
AK088867.1 | - | 2 - 3 | 363 | 53 | |
AK171507.1 | - | 2 - 3 | 387 | 53 | |
AY255786.1 | - | 1 - 2 | 337 | 53 | |
AY255787.1 | - | 1 - 2 | 337 | 53 | |
BC011067.1 | - | 2 - 3 | 353 | 53 | |
BC024859.1 | - | 1 - 2 | 1581 | 53 | |
CN716634.1 | - | 2 - 3 | 440 | 53 |