Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020028.3 | NP_064412.2 | 2 - 3 | 887 | 116 |
GenBank mRNA | AK051092.1 | - | 2 - 3 | 837 | 116 |
AK144861.1 | - | 2 - 3 | 888 | 116 | |
BC060131.1 | - | 2 - 3 | 1023 | 116 | |
BC064676.1 | - | 2 - 3 | 1023 | 116 |