Assay Design



Gene Transcripts

Gene Symbol
Erbb2ip
Entrez Gene ID
59079
Gene Name
Erbb2 interacting protein
Gene Aliases
1700028E05Rik, Erbin, mKIAA1225
Chromosome Location
Chr.13: 103818786 - 103920586 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001289473.1 NP_001276402.1 21 - 22 4007 64
GenBank mRNA AF263743.1 - 1 - 2 888 64
AK051733.1 - 1 - 2 666 64
AK122473.1 - 21 - 22 3686 64
BC005691.1 - 1 - 2 132 64
BC145509.1 - 21 - 22 3716 64

View Full Product Details