Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023547.1 | NP_076036.1 | 3 - 4 | 332 | 90 |
GenBank mRNA | AK010946.1 | - | 3 - 4 | 434 | 90 |
BC019383.1 | - | 3 - 4 | 366 | 90 | |
BC047990.1 | - | 3 - 4 | 367 | 90 | |
BC058788.1 | - | 3 - 4 | 378 | 90 |