Assay Design



Gene Transcripts

Gene Symbol
Esm1
Entrez Gene ID
71690
Gene Name
endothelial cell-specific molecule 1
Gene Aliases
0610042H23Rik, AV004503, ESM-1
Chromosome Location
Chr.13: 113209659 - 113218104 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023612.3 NP_076101.1 1 - 2 489 69
GenBank mRNA AJ249354.1 - 1 - 2 352 69
AK002913.1 - 1 - 2 383 69
BC051919.1 - 1 - 2 489 69
BC147648.1 - 1 - 2 357 69
BC147650.1 - 1 - 2 357 69

View Full Product Details