Assay Design



Gene Transcripts

Gene Symbol
Yipf7
Entrez Gene ID
75581
Gene Name
Yip1 domain family, member 7
Gene Aliases
2310016N21Rik, Yip1b
Chromosome Location
Chr.5: 69516670 - 69542647 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023784.5 NP_076273.1 5 - 6 693 63
GenBank mRNA AF217188.1 - 4 - 5 605 63
AK009405.1 - 5 - 6 644 63
BC089576.1 - 5 - 6 678 63

View Full Product Details