Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023831.3 | NP_076320.2 | 4 - 5 | 476 | 72 |
GenBank mRNA | AJ249981.1 | - | 4 - 5 | 520 | 72 |
AK005360.1 | - | 4 - 5 | 484 | 72 | |
AK032880.1 | - | 4 - 5 | 477 | 72 | |
BC010326.1 | - | 3 - 4 | 275 | 72 | |
BC080764.1 | - | 4 - 5 | 418 | 72 |