Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023844.5 | NP_076333.3 | 1 - 2 | 427 | 77 |
GenBank mRNA | AF255911.1 | - | 1 - 2 | 150 | 77 |
AJ291757.1 | - | 1 - 2 | 363 | 77 | |
AK010616.1 | - | 1 - 2 | 269 | 77 | |
AK010826.1 | - | 1 - 2 | 271 | 77 | |
AK013914.1 | - | 1 - 2 | 268 | 77 | |
AK028757.1 | - | 1 - 2 | 118 | 77 | |
AK045095.1 | - | 1 - 2 | 165 | 77 | |
AK078128.1 | - | 1 - 2 | 153 | 77 | |
BC028778.1 | - | 1 - 2 | 427 | 77 |