Assay Design



Gene Transcripts

Gene Symbol
Zfpl1
Entrez Gene ID
81909
Gene Name
zinc finger like protein 1
Gene Aliases
1500015B20Rik
Chromosome Location
Chr.19: 6077366 - 6084939 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024231.2 NP_077193.1 7 - 8 786 153
GenBank mRNA AK005244.1 - 7 - 8 798 153
BC002119.1 - 7 - 8 786 153

View Full Product Details