Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026238.4 | NP_080514.3 | 6 - 7 | 734 | 77 |
GenBank mRNA | AK003464.1 | - | 1 - 2 | 67 | 77 |
AK014038.1 | - | 6 - 7 | 718 | 77 | |
AK018548.1 | - | 6 - 7 | 705 | 77 | |
AK043694.1 | - | 6 - 7 | 711 | 77 | |
AK145408.1 | - | 6 - 7 | 737 | 77 | |
AK187867.1 | - | 3 - 4 | 334 | 77 | |
BC020468.1 | - | 6 - 7 | 694 | 77 | |
BC025108.1 | - | 6 - 7 | 714 | 77 | |
BC052830.1 | - | 6 - 7 | 711 | 77 |