Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026318.3 | NP_080594.2 | 3 - 4 | 314 | 71 |
GenBank mRNA | AI847440.1 | - | 1 - 2 | 71 | |
AK003580.1 | - | 3 - 4 | 308 | 71 | |
AK009109.1 | - | 3 - 4 | 302 | 71 | |
AK168506.1 | - | 3 - 4 | 283 | 71 | |
BC021588.1 | - | 2 - 3 | 568 | 71 | |
BC038469.1 | - | 3 - 4 | 316 | 71 | |
BC060244.1 | - | 2 - 3 | 1063 | 71 | |
BY165803.1 | - | 3 - 4 | 316 | 71 |