Assay Design



Gene Transcripts

Gene Symbol
Ift74
Entrez Gene ID
67694
Gene Name
intraflagellar transport 74
Gene Aliases
1700029H06Rik, Ccdc2, Cmg1, b2b796Clo
Chromosome Location
Chr.4: 94603266 - 94693233 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026319.3 NP_080595.2 18 - 19 1802 64
GenBank mRNA AK006492.1 - 9 - 10 909 64
CD740935.1 - 2 - 3 64

View Full Product Details