Assay Design



Gene Transcripts

Gene Symbol
Slc25a37
Entrez Gene ID
67712
Gene Name
solute carrier family 25, member 37
Gene Aliases
1700020E22Rik, 4930513O14Rik, 4930526G11Rik, AI848481, C330015G08Rik, Mfrn, Mfrn1, Mscp, frascati, mitoferrin
Chromosome Location
Chr.14: 69241848 - 69285103 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026331.3 NP_080607.2 2 - 3 600 61
GenBank mRNA AF288621.1 - 2 - 3 553 61
AK015770.1 - 2 - 3 300 61
AK050414.1 - 2 - 3 568 61

View Full Product Details