Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001026211.2 | NP_001021382.1 | 2 - 3 | 221 | 76 |
NM_001286972.1 | NP_001273901.1 | 2 - 3 | 221 | 76 | |
NM_026332.4 | NP_080608.3 | 2 - 3 | 221 | 76 | |
GenBank mRNA | AK007866.1 | - | 2 - 3 | 189 | 76 |
AK008267.1 | - | 2 - 3 | 165 | 76 | |
AK010204.1 | - | 2 - 3 | 171 | 76 | |
AK012051.1 | - | 2 - 3 | 173 | 76 | |
AK076602.1 | - | - | 180 | 76 | |
BC020327.1 | - | 2 - 3 | 154 | 76 | |
BC024335.1 | - | 2 - 3 | 129 | 76 | |
BY149679.1 | - | 2 - 3 | 194 | 76 | |
CA469052.1 | - | 2 - 3 | 258 | 76 | |
EL962106.1 | - | 2 - 3 | 167 | 76 |