Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142681.1 | NP_001136153.1 | 5 - 6 | 569 | 91 |
NM_026522.5 | NP_080798.2 | 5 - 6 | 569 | 91 | |
GenBank mRNA | AK014071.1 | - | 5 - 6 | 518 | 91 |
AK014141.1 | - | 5 - 6 | 544 | 91 | |
AK050655.1 | - | 5 - 6 | 517 | 91 | |
AK147377.1 | - | 5 - 6 | 532 | 91 | |
AK160156.1 | - | 5 - 6 | 529 | 91 | |
AK160200.1 | - | 5 - 6 | 529 | 91 | |
BC006876.1 | - | 5 - 6 | 518 | 91 | |
BC061063.1 | - | 5 - 6 | 529 | 91 |