Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026598.3 | NP_080874.2 | 1 - 2 | 224 | 87 |
GenBank mRNA | AF243434.2 | - | 1 - 2 | 180 | 87 |
AK011237.1 | - | 1 - 2 | 183 | 87 | |
AK019140.1 | - | 1 - 2 | 76 | 87 | |
AK019963.1 | - | 1 - 2 | 219 | 87 | |
AK032534.1 | - | 1 - 2 | 188 | 87 | |
AK163371.1 | - | 1 - 2 | 183 | 87 | |
BC027422.1 | - | 1 - 2 | 202 | 87 | |
BY349880.1 | - | 1 - 2 | 227 | 87 |