Assay Design



Gene Transcripts

Gene Symbol
Odf3
Entrez Gene ID
69287
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
1700011O04Rik, SHIPPO1
Chromosome Location
Chr.7: 140847916 - 140850925 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027019.3 NP_081295.2 2 - 3 138 61
GenBank mRNA AB067773.1 - 2 - 3 227 61
AK005883.1 - 2 - 3 249 61
BC104105.1 - 2 - 3 138 61
BC104106.1 - 2 - 3 138 61
BC106806.1 - 2 - 3 138 61

View Full Product Details