Assay Design



Gene Transcripts

Gene Symbol
Msh4
Entrez Gene ID
55993
Gene Name
mutS homolog 4 (E. coli)
Gene Aliases
4930485C04Rik, AV144863, mMsh4
Chromosome Location
Chr.3: 153857141 - 153906138 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282054.1 NP_001268983.1 12 - 13 1669 104
NM_031870.3 NP_114076.1 11 - 12 1760 104
GenBank mRNA AF178957.1 - 11 - 12 1464 104
AF298655.1 - 11 - 12 1760 104
AK029837.1 - 12 - 13 1670 104
AK161474.1 - 11 - 12 1510 104
AY351585.1 - 11 - 12 1714 104
AY351590.1 - 3 - 4 1101 104
AY351591.1 - 3 - 4 1101 104
BC138163.1 - 11 - 12 1651 104
BC145838.1 - 11 - 12 1651 104

View Full Product Details