Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033072.2 | NP_149063.2 | 11 - 12 | 3035 | 50 |
GenBank mRNA | AK018262.1 | - | 2 - 3 | 176 | 50 |
AK047331.1 | - | 1 - 2 | 60 | 50 | |
AK158797.1 | - | 11 - 12 | 3153 | 50 | |
BC019433.1 | - | 3 - 4 | 536 | 50 | |
BC038259.1 | - | 2 - 3 | 212 | 50 | |
BC049833.1 | - | 3 - 4 | 519 | 50 | |
BC055445.1 | - | 3 - 4 | 492 | 50 | |
BC083328.1 | - | 2 - 3 | 172 | 50 | |
BC096027.1 | - | 8 - 9 | 2777 | 50 |