Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159986.1 | NP_001153458.1 | 15 - 16 | 1975 | 60 |
NM_033354.3 | NP_203505.3 | 17 - 18 | 2222 | 60 | |
NR_027641.1 | 16 - 17 | 2054 | 60 | ||
GenBank mRNA | AB063356.1 | - | 15 - 16 | 1975 | 60 |
AK132144.1 | - | 15 - 16 | 1999 | 60 | |
AK173302.1 | - | 17 - 18 | 2203 | 60 | |
BC039223.1 | - | 7 - 8 | 826 | 60 | |
BC051110.1 | - | 8 - 9 | 922 | 60 | |
BC059194.1 | - | 16 - 17 | 2040 | 60 |