Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159986.1 | NP_001153458.1 | 17 - 18 | 2226 | 70 |
NM_033354.3 | NP_203505.3 | 19 - 20 | 2473 | 70 | |
NR_027641.1 | 18 - 19 | 2305 | 70 | ||
GenBank mRNA | AB063356.1 | - | 17 - 18 | 2226 | 70 |
AK132144.1 | - | 17 - 18 | 2250 | 70 | |
AK173302.1 | - | 19 - 20 | 2454 | 70 | |
BC039223.1 | - | 9 - 10 | 1077 | 70 | |
BC051110.1 | - | 10 - 11 | 1173 | 70 | |
BC059194.1 | - | 18 - 19 | 2291 | 70 |