Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272028.1 | NP_001258957.1 | 6 - 7 | 1900 | 64 |
NM_001272029.1 | NP_001258958.1 | 7 - 8 | 2051 | 64 | |
NM_001272030.1 | NP_001258959.1 | 6 - 7 | 1900 | 64 | |
NM_001272031.1 | NP_001258960.1 | 5 - 6 | 1643 | 64 | |
NM_001272032.1 | NP_001258961.1 | 5 - 6 | 1643 | 64 | |
NM_033474.3 | NP_258435.2 | 7 - 8 | 2051 | 64 | |
GenBank mRNA | AF286212.1 | - | 6 - 7 | 1900 | 64 |
AF286213.1 | - | 6 - 7 | 1900 | 64 | |
AF286214.1 | - | 5 - 6 | 1552 | 64 | |
AF286215.1 | - | 5 - 6 | 1552 | 64 | |
AJ243418.1 | - | 6 - 7 | 1685 | 64 | |
AK044982.1 | - | 5 - 6 | 1591 | 64 | |
AK051627.1 | - | 5 - 6 | 1599 | 64 | |
BC052393.1 | - | 7 - 8 | 1970 | 64 | |
BC056980.1 | - | 7 - 8 | 1959 | 64 |