Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272028.1 | NP_001258957.1 | 9 - 10 | 2160 | 65 |
NM_001272029.1 | NP_001258958.1 | 9 - 10 | 2293 | 65 | |
NM_001272030.1 | NP_001258959.1 | 8 - 9 | 2142 | 65 | |
NM_001272031.1 | NP_001258960.1 | 8 - 9 | 1903 | 65 | |
NM_001272032.1 | NP_001258961.1 | 7 - 8 | 1885 | 65 | |
NM_033474.3 | NP_258435.2 | 10 - 11 | 2311 | 65 | |
GenBank mRNA | AF286212.1 | - | 9 - 10 | 2160 | 65 |
AF286213.1 | - | 8 - 9 | 2142 | 65 | |
AF286214.1 | - | 8 - 9 | 1812 | 65 | |
AF286215.1 | - | 7 - 8 | 1794 | 65 | |
AJ243418.1 | - | 8 - 9 | 1927 | 65 | |
AK031855.1 | - | 3 - 4 | 268 | 65 | |
AK044982.1 | - | 7 - 8 | 1833 | 65 | |
AK051627.1 | - | 8 - 9 | 1859 | 65 | |
AK084886.1 | - | 9 - 10 | 2281 | 65 | |
BC051655.1 | - | 4 - 5 | 308 | 65 | |
BC052393.1 | - | 9 - 10 | 2212 | 65 | |
BC056980.1 | - | 10 - 11 | 2219 | 65 |