Assay Design



Gene Transcripts

Gene Symbol
Nrep
Entrez Gene ID
27528
Gene Name
neuronal regeneration related protein
Gene Aliases
AI325076, D0H4S114, Harp, P311, PTZ17, SEZ17
Chromosome Location
Chr.18: 33437019 - 33464029 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109988.1 NP_001103458.1 2 - 3 221 85
GenBank mRNA AK020629.1 - 2 - 3 82 85
CJ165393.1 - 2 - 3 224 85

View Full Product Details