Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109988.1 | NP_001103458.1 | 4 - 5 | 360 | 91 |
NM_001109989.2 | NP_001103459.1 | 4 - 5 | 283 | 91 | |
NM_001267717.1 | NP_001254646.1 | 4 - 5 | 272 | 91 | |
NM_053078.4 | NP_444308.1 | 3 - 4 | 398 | 91 | |
GenBank mRNA | AK020629.1 | - | 4 - 5 | 221 | 91 |
AK048361.1 | - | 3 - 4 | 198 | 91 | |
AK081502.1 | - | 3 - 4 | 199 | 91 | |
AK133769.1 | - | 2 - 3 | 1493 | 91 | |
BC054762.1 | - | 3 - 4 | 180 | 91 | |
BQ715224.1 | - | 3 - 4 | 185 | 91 | |
BU057200.1 | - | 3 - 4 | 188 | 91 | |
CJ148521.1 | - | 3 - 4 | 503 | 91 | |
CX768079.1 | - | 4 - 5 | 281 | 91 | |
EU447302.1 | - | 2 - 3 | 126 | 91 | |
X70398.1 | - | 3 - 4 | 194 | 91 |