Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195031.1 | NP_001181960.1 | 6 - 7 | 956 | 68 |
NM_053182.5 | NP_444412.2 | 5 - 6 | 897 | 68 | |
GenBank mRNA | AF250192.1 | - | 3 - 4 | 278 | 68 |
AK040230.1 | - | 6 - 7 | 906 | 68 | |
AK076515.1 | - | 2 - 3 | 395 | 68 | |
AK089329.1 | - | 5 - 6 | 849 | 68 | |
AK156000.1 | - | 6 - 7 | 908 | 68 | |
AK156986.1 | - | 6 - 7 | 881 | 68 | |
AK172106.1 | - | 6 - 7 | 879 | 68 | |
BC145328.1 | - | 6 - 7 | 780 | 68 | |
BC145761.1 | - | 6 - 7 | 780 | 68 | |
BC145763.1 | - | 6 - 7 | 780 | 68 |