Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053244.5 | NP_444474.1 | 4 - 5 | 2443 | 81 |
GenBank mRNA | AF343726.1 | - | 4 - 5 | 740 | 81 |
AK039628.1 | - | 4 - 5 | 2444 | 81 | |
AK139303.1 | - | 2 - 3 | 3137 | 81 | |
AY044228.1 | - | 4 - 5 | 773 | 81 | |
BC016531.1 | - | 4 - 5 | 886 | 81 | |
EU532437.1 | - | 4 - 5 | 740 | 81 |