Assay Design
Gene Transcripts
- Gene Symbol
- Foxp3
- Entrez Gene ID
-
20371
- Gene Name
- forkhead box P3
- Gene Aliases
-
JM2, scurfin, sf
- Chromosome Location
-
Chr.X: 7579676 - 7595243 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_054039.2 |
NP_473380.1 |
1 - 2 |
171 |
72 |
GenBank mRNA |
AF277992.1 |
- |
1 - 2 |
154 |
72 |
View Full Product Details