Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199347.1 | NP_001186276.1 | 3 - 4 | 326 | 87 |
NM_001199348.1 | NP_001186277.1 | 2 - 3 | 184 | 87 | |
NM_054039.2 | NP_473380.1 | 2 - 3 | 259 | 87 | |
GenBank mRNA | AF277991.1 | - | 2 - 3 | 184 | 87 |
AF277992.1 | - | 2 - 3 | 242 | 87 | |
AK157447.1 | - | 3 - 4 | 327 | 87 | |
DQ387959.1 | - | 3 - 4 | 326 | 87 |