Assay Design
Gene Transcripts
- Gene Symbol
- Brdt
- Entrez Gene ID
-
114642
- Gene Name
- bromodomain, testis-specific
- Gene Aliases
-
7420412D09Rik, Brd6, Fsrg3
- Chromosome Location
-
Chr.5: 107331091 - 107387061 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_054054.2 |
NP_473395.2 |
8 - 9 |
1395 |
77 |
GenBank mRNA |
AF358660.1 |
- |
8 - 9 |
1430 |
77 |
View Full Product Details